Skip to Content
Merck
All Photos(2)

Key Documents

5.32605

Sigma-Aldrich

Aurora A/MYCN Dual Inhibitor, CD532

Synonym(s):

Aurora A/MYCN Dual Inhibitor, CD532, Aurora A Inhibitor, CD532, MYCN Inhibitor, CD532

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€190.00

€190.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€190.00

About This Item

Empirical Formula (Hill Notation):
C26H25F3N8O
CAS Number:
Molecular Weight:
522.52
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€190.00


Estimated to ship on21 May 2025


Request a Bulk Order

Assay

≥97% (HPLC)

Quality Level

form

solid

potency

48 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

tan

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=CC(C(F)(F)F)=C1)NC2=CC=C(NC3=NC=CC(NC4=NNC(C5CCCC5)=C4)=N3)C=C2

InChI key

GBMIFBVLJSCVJT-UHFFFAOYSA-N

General description

A cell permeable diaminopyrimidine derived compound that acts as a highly potent and selective inhibitor of Aurora A (IC50 = 48 nM). Acts by binding to the hinge region via a pyrazole moiety and stabilizes a DFG-in, inactive conformation of Aurora A. Potentiates the loss of the wild-type MYCN protein by disrupting MYCN-Aurora A complex and allowing its ubiquitination and proteasomal degradation in MYCN-amplified SK-N-BE (2) and Kelly neuroblastoma cells (EC50 = 223 and 146.7 nM, respectively). Causes a loss of S-phase entry of cells and allows their accumulation in both G0/G1 and G2 phases. Shown to be effective in vivo with serum t1/2 = 1.5 h in mice.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Aurora A
Reversible: yes
Secondary Target
MYCN

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lee, J.K., et al. 2016. Cancer Cell.29, In press.
Gustafson, W. C., et al. 2014. Cancer Cell.26, 414.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service