Skip to Content
Merck
All Photos(1)

Key Documents

5.31170

Sigma-Aldrich

ALDH3A1 Inhibitor, CB29

Synonym(s):

ALDH3A1 Inhibitor, CB29, Aldehyde dehydrogenase ALDH3A1 inhibitor, CB29

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€235.00

€235.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€235.00

About This Item

Empirical Formula (Hill Notation):
C15H15N3O5S
Molecular Weight:
349.36
UNSPSC Code:
12352200
NACRES:
NA.77

€235.00


Estimated to ship on28 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

powder

potency

4.7 μM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell permeable diarylamine derivative that acts as a selective, reversible, substrate-binding site directed inhibitor of aldehyde dehydrogenase ALDH3A1 (Ki = 4.7 µM, IC50 = 16 µM). Does not inhibit the activity of ALDH1A1, ALDH1A2, ALDH1A3, ALDH1B1, and ALDH2 even at higher concentrations (250 µM). Shown to significantly reduce the activity of ALDH3A1 in A549 and SF767 cell lysates, but does not affect CCD-13Lu cells that lack ALDH3A1 activity. Although it does not affect tumor cell proliferation, it significantly improves the sensitivity of cancer cells to mafosfamide.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Primary Target
ALDH3A1
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Parajuli, B. et al. 2014. Chembiochem.15, 701.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service