Skip to Content
Merck
All Photos(1)

Key Documents

5.31085

Sigma-Aldrich

Eya2 Phosphatase Inhibitor, MLS000544460

Synonym(s):

Eya2 Phosphatase Inhibitor, MLS000544460, 3-Fluoro-N-(( E)-(5-pyridin-2-ylsulfanylfuran-2-yl)methylideneamino)benzamide, ( E)-3-Fluoro-Nʹ-((5-(pyridin-2-ylthio)furan-2-yl)methylene)benzohydrazide, Eab1 Inhibitor, Eyes Absent Homolog 2 Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€207.00

€207.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€207.00

About This Item

Empirical Formula (Hill Notation):
C17H12FN3O2S
CAS Number:
Molecular Weight:
341.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€207.00


Estimated to ship on21 May 2025


Request a Bulk Order

Assay

≥97% (HPLC)

Quality Level

form

solid

potency

2.0 μM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable furylidenebenzohydrazide compound that acts as a reversible inhibitor against Eya2 C-terminal Eya domain (ED) protein tyrosine phosphatase (PTP) activity (IC50 = 4.1 µM; 100 nM ED & 5 µM Mg2+; Substrate = 25 µM OMFP) via affinity docking at an allosteric pocket located opposite to the active site (KD = 6.1 or 2.0 µM, respectively, with or without 5 mM exogenous Mg2+; KD = 0.8 µM with 10 mM EDTA and no exogenous Mg2+), while being ineffective against the PTP activity of Eya3 & PTP1B or the Ser/Thr phosphatase activity of PPM1A & Scp1. Shown to reduce Eya2 overexpression-induced migration enhancement in MCF10A cultures (17% vs. 54% enhancement with or without 10 µM inhibitor). The inhibition appears to be non-competitive with respect to substrate in the presence of 5 mM Mg2+, while mixed mode of inhibition is observed at a low Mg2+ concentration of 5 µM.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Eya2 C-terminal Eya domain (ED) protein tyrosine phosphatase
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Krueger, A.B., et al. 2014. J. Biol. Chem.289, 16349.

Krueger, A.B., et al. 2013. J. Biomol. Screen.18, 85.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Konstantinos Sousounis et al.
eLife, 9 (2020-03-07)
How salamanders accomplish progenitor cell proliferation while faithfully maintaining genomic integrity and regenerative potential remains elusive. Here we found an innate DNA damage response mechanism that is evident during blastema proliferation (early- to late-bud) and studied its role during tissue

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service