Skip to Content
Merck
All Photos(1)

Key Documents

5.30961

Sigma-Aldrich

TAZ Activator, IBS008738

Synonym(s):

TAZ Activator, IBS008738, (4-((morpholinomethylene)amino)-1-(p-tolyl)-1H-imidazol-5-yl)(phenyl)methanone, Transcriptional Co-activator with PDZ-binding Motif (TAZ) Activator, WWTR1 Activator

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€160.00

€160.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€160.00

About This Item

Empirical Formula (Hill Notation):
C22H22N4O2
CAS Number:
Molecular Weight:
374.44
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€160.00


Estimated to ship on30 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

SMILES string

CC1=CC=C(C=C1)N2C=NC(=C2C(=O)C3=CC=CC=C3)N=CN4CCOCC4

InChI

1S/C22H22N4O2/c1-17-7-9-19(10-8-17)26-16-24-22(23-15-25-11-13-28-14-12-25)20(26)21(27)18-5-3-2-4-6-18/h2-10,15-16H,11-14H2,1H3

InChI key

UCDKLMTYEDPEMY-UHFFFAOYSA-N

General description

A benzoylimidazole derivative that acts as a Transcriptional co-activator with PDZ-binding motif (TAZ) Activator by stabilizing TAZ and increases unphosphorylated TAZ. Up-regulates TAZ-dependent MyoD-dependent gene transcription at 10 µM and competes with myostatin in C2C12 cells. Shown to enhance myogenesis most effectively when applied for the first 24 hours under the differentiation condition. Facilitates muscle repair in cardiotoxin-induced muscle injury and prevents dexamethasone-induced muscle atrophy.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Yang, Z., et al. 2014. Mol. Cell. Biol.34, 1607.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service