Skip to Content
Merck
All Photos(2)

Documents

5.30959

Sigma-Aldrich

TAZ Activity Modulator, TM-25659

Synonym(s):

TAZ Activity Modulator, TM-25659, TM25659, TM 25659

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H28N8
CAS Number:
Molecular Weight:
500.60
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

CC1=NC2=C(N=C(CCCC)N2CC3=CC=C(C4=CC=CC=C4C5=NN=NN5)C=C3)C=C1C6=CN=CC=C6

General description

A cell permeable, orally bioavailable imidazol-[4,5-b]pyridine derivative that enhances nuclear localization of transcriptional co-activator with PDZ-binding motif (TAZ) in a dose-dependent manner without affecting the total amount of TAZ in pluripotent C3H10T1/2 cells. Does not affect Ser89 phosphorylation in TAZ, but reduces tyrosine phosphorylation. Reduces PPARg levels in differentiated adipocytes and acts as a suppressor of PPARg-dependent adipocyte differentiation. Also shown to enhance RUNX2-induced osteoblast differentiation of C3H10T1/2 cells and mineralization in a dose-dependent manner. Reduces weight gain in ob/ob mice (50 mg/kg, i.p.) and attenuates bone loss in ovariectomized mice. Displays desirable pharmacokinetic properties with t1/2 = 9.85 h.

Please note that the molecular weight for this compound is batch-specific due to variable water content.
A cell permeable, orally bioavailable imidazol-pyrimidine derivative that enhances nuclear localization of transcriptional co-activator with PDZ-binding motif (TAZ) in a dose-dependent manner without affecting the total amount of TAZ in pluripotent C3H10T1/2 cells. Does not affect Ser89 phosphorylation in TAZ, but reduces tyrosine phosphorylation. Reduces PPARg levels in differentiated adipocytes and acts as a suppressor of PPARg-dependent adipocyte differentiation. Also shown to enhance RUNX2-induced osteoblast differentiation of C3H10T1/2 cells and mineralization in a dose-dependent manner. Reduces weight gain in ob/ob mice (50 mg/kg, i.p.) and attenuates bone loss in ovariectomized mice. Displays desirable pharmacokinetic properties with t1/2 = 9.85 h.
A cell-permeable enhancer of nuclear localization of transcriptional co-activator with PDZ-binding motif (TAZ). Does not affect the total amount of TAZ in pluripotent C3H10T1/2 cells.

Biochem/physiol Actions

Cell permeable: yes

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Jang, E.J., et al. 2012. Br. J. Pharmacol.165, 1584.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service