Skip to Content
Merck
All Photos(1)

Key Documents

5.30509

Sigma-Aldrich

StemSelect Compound E

≥95% (HPLC), liquid, γ-secretase inhibitor, Calbiochem®

Synonym(s):

StemSelect Compound E, ( S, S)- 2-(2-(3,5-Difluorophenyl)-acetylamino)-N-(1-methyl-2-oxo-5-phenyl-2,3-dihydro-1H-benzo[e][1,4]diazepin-3-yl)-propionamide, InSolution γ-Secretase Inhibitor XXI, Compound E

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
€214.00

€214.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
250 μG
€214.00

About This Item

Empirical Formula (Hill Notation):
C27H24F2N4O3
CAS Number:
Molecular Weight:
490.50
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€214.00


Estimated to ship on28 May 2025


Request a Bulk Order

Product Name

StemSelect Compound E,

Assay

≥95% (HPLC)

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

SMILES string

Fc1cc(cc(c1)CC(=O)N[C@@H](C)C(=O)NC2N=C(c4c(cccc4)N(C2=O)C)c3ccccc3)F

InChI

1S/C27H24F2N4O3/c1-16(30-23(34)14-17-12-19(28)15-20(29)13-17)26(35)32-25-27(36)33(2)22-11-7-6-10-21(22)24(31-25)18-8-4-3-5-9-18/h3-13,15-16,25H,14H2,1-2H3,(H,30,34)(H,32,35)/t16-,25?/m0/s1

InChI key

JNGZXGGOCLZBFB-YPHZTSLFSA-N

General description

Notch signaling suppressor.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
presenilins

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

A 5 mM (250 µg/102 µl), 0.2 µm-filtered solution in DMSO.

Reconstitution

Following initial use, aliquot and freeze (-70°C).

Other Notes

Li, W., et al. 2011. Proc. Natl Acad. Sci. USA108, 8299.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service