Skip to Content
Merck
All Photos(1)

Key Documents

529606

Sigma-Aldrich

PP1 Analog II, 1NM-PP1

InSolution, ≥95%, reversible PP1 analog that acts as a potent, selective, and ATP-competitive inhibitor of mutant kinases

Synonym(s):

InSolution PP1 Analog II, 1NM-PP1, 4-Amino-1- tert-butyl-3-(1ʹ-naphthylmethyl)pyrazolo[3,4-d]pyrimidine, 4-Amino-1-tert-butyl-3-(1ʹ-naphthylmethyl)pyrazolo[3,4-d]pyrimidine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€176.00

€176.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€176.00

About This Item

Empirical Formula (Hill Notation):
C20H21N5
CAS Number:
Molecular Weight:
331.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€176.00


Estimated to ship on26 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

shipped in

dry ice

storage temp.

−70°C

SMILES string

[n]1(nc(c4c1ncnc4N)Cc2c3c(ccc2)cccc3)C(C)(C)C

InChI

1S/C20H21N5/c1-20(2,3)25-19-17(18(21)22-12-23-19)16(24-25)11-14-9-6-8-13-7-4-5-10-15(13)14/h4-10,12H,11H2,1-3H3,(H2,21,22,23)

InChI key

GDQXJQSQYMMKRA-UHFFFAOYSA-N

General description

A cell-permeable and reversible PP1 analog that acts as a potent, selective, and ATP-competitive inhibitor of mutant kinases compared to the corresponding wild-type kinase (IC50 = 3.2 nM for T339G, c-Fyn-as1 vs. 1.0 µM for c-Fyn; 4.3 nM for I338G, v-src-as1 vs. 28 µM for v-src; 5 nM for F80G, CDK2-as1 vs. 29 µM for CDK2; 8 nM for F89G, CAMK IIa-as1 vs. 24 µM for CAMKII; 120 nM for T315A, c-Abl-as2 vs. 3.4 µM for c-Abl). Shown to activate mutants of Ire1, a transmembrane kinase.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
mutant c-Fyn, v-src, Cadk2, CaMKIIa, c-Abl etc.,
Reversible: yes
Target IC50: 3.2 nM for T339G, c-Fyn-as1 vs. 1.0 &micro

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (1 mg/302 µL) solution of PP1 Analog II, 1NM-PP1 (Cat. No. 529581) in DMSO.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Aliquots are stable for up to 3 months at -70°C.

Other Notes

Ira, G., et al. 2004. Nature431, 1011.
Papa, F.R., et al. 2003. Science302, 1533.
Bishop, A.C., et al. 2000. Nature407, 395.
Weiss, E.L., et al. 2000. Nat. Cell Biol.2, 677.
Bishop, A.C., et al. 1999. J. Am. Chem. Soc.121, 627.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup - Information refers to the main ingredient.

Flash Point(C)

87 °C - closed cup - Information refers to the main ingredient.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service