Skip to Content
Merck
All Photos(1)

Key Documents

529567-M

Millipore

Potassium Phosphate, Dibasic, Trihydrate, Molecular Biology Grade

Synonym(s):

Potassium Phosphate, Dibasic, Trihydrate, Molecular Biology Grade, K₂HPO₄

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€248.00

€248.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
1 KG
€248.00

About This Item

Empirical Formula (Hill Notation):
HK2O4P · 3H2O
CAS Number:
Molecular Weight:
228.22
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.25

€248.00


Estimated to ship on24 May 2025


Request a Bulk Order

grade

Molecular Biology

Quality Level

description

Merck USA index - 14, 7658

Assay

≥98% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

pKa (20 °C)

7.21

solubility

water: soluble

cation traces

heavy metals: ≤0.001%

absorption

≤0.01 at 260 nm
≤0.01 at 280 nm

foreign activity

DNases, proteases, RNases, none detected

storage temp.

10-30°C

SMILES string

[K+].[K+].[P](=O)([O-])([O-])O.O.O.O

InChI

1S/2K.H3O4P.3H2O/c;;1-5(2,3)4;;;/h;;(H3,1,2,3,4);3*1H2/q2*+1;;;;/p-2

InChI key

XQGPKZUNMMFTAL-UHFFFAOYSA-L

General description

Biological buffer (absorbance: ≤0.01 at 260 nm and 280 nm).

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service