Skip to Content
Merck

528113

PI 3-Kβ Inhibitor VI, TGX-221 - CAS 663619-89-4 - Calbiochem

The PI 3-Kβ Inhibitor VI, TGX-221, also referenced under CAS 663619-89-4, controls the biological activity of PI 3-Kβ. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

PI 3-Kβ Inhibitor VI, TGX-221 - CAS 663619-89-4 - Calbiochem, (±)-7-Methyl-2-(morpholin-4-yl)-9-(1-phenylaminoethyl)-pyrido[1,2-a]-pyrimidin-4-one, PI 3-K Inhibitor VI

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H24N4O2
CAS Number:
Molecular Weight:
364.44
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

white

solubility

DMSO: 10 mg/mL, ethanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable morpholino-pyrimidinone compound that acts as a reversible, ATP-competitive, selective, and highly potent inhibitor of PI 3-Kβ (IC50 = 0.005, 0.1, 5, and ≥3.5 µM for -β, -δ, -α and -γ isoforms, respectively) with little activity against a panel of 15 commonly studied protein kinases even at concentrations as high as 10.0 µM. An excellent tool for studying p110β-dependent responses both in cells in vitro and in animals in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PI 3-Kβ
Product competes with ATP.
Reversible: yes
Target IC50: 0.005, 0.1, 5, and ≥3.5 µM for -β, -δ, -α and -γ isoforms, respectively

Packaging

Packaged under inert gas

Other Notes

Chaussade, C., et al. 2007. Biochem. J.404, 449.
Cosemans, J.M, et al. 2006. Blood108, 3045.
Condliffe, A.M., et al. 2005. Blood106, 1432.
Jackson, S.P., et al. 2005. Nat. Med.11, 507.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library