Skip to Content
Merck
All Photos(2)

Key Documents

527451

Sigma-Aldrich

PKR Inhibitor

InSolution, ≥90%

Synonym(s):

InSolution PKR Inhibitor, Double-stranded RNA-activated Protein Kinase Inhibitor I, Double-stranded RNA-dependent Protein Kinase Inhibitor I, EIF2AK2 Inhibitor I, C16

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€190.00

€190.00


Estimated to ship on23 May 2025



Select a Size

Change View
5 MG
€190.00

About This Item

Empirical Formula (Hill Notation):
C13H8N4OS
CAS Number:
Molecular Weight:
268.29
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€190.00


Estimated to ship on23 May 2025


Quality Level

Assay

≥90% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

shipped in

dry ice

storage temp.

−20°C

SMILES string

[s]1c2c(nc1)ccc3c2\C(=C\c4nc[nH]c4)\C(=O)N3

InChI

1S/C13H8N4OS/c18-13-8(3-7-4-14-5-15-7)11-9(17-13)1-2-10-12(11)19-6-16-10/h1-6H,(H,14,15)(H,17,18)/b8-3-

InChI key

VFBGXTUGODTSPK-BAQGIRSFSA-N

General description

An imidazolo-oxindole compound that acts as a potent, ATP-binding site directed inhibitor of PKR. Shown to effectively inhibit RNA-induced PKR autophosphorylation (IC50 = 210 nM) and rescue PKR-dependent translation block (IC50 = 100 nM). The solid form of this compound (Cat. No. 527450 ) is also available.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKR
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 50 mM (5 mg/373 µL) solution of PKR Inhibitor (Cat. No. 527450 ) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Aliquots are stable for up to 6 months at -20°C.

Other Notes

Zhu, P.J., et al. 2011. Cell147, 1384.
Jammi, N.V., et al. 2003. Biochem. Biophys. Res. Commun.308, 50.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Narasimham V Jammi et al.
Biochemical and biophysical research communications, 308(1), 50-57 (2003-08-02)
The RNA-dependent protein kinase (PKR) is an interferon-induced serine/threonine protein kinase that phosphorylates the alpha subunit of the eukaryotic initiation factor 2 in response to viral infection. Classical genetic approaches for studying the role of PKR in cell signaling have
Ping Jun Zhu et al.
Cell, 147(6), 1384-1396 (2011-12-14)
The double-stranded RNA-activated protein kinase (PKR) was originally identified as a sensor of virus infection, but its function in the brain remains unknown. Here, we report that the lack of PKR enhances learning and memory in several behavioral tasks while
Harikrishnan Radhakrishnan et al.
Bioengineering & translational medicine, 9(1), e10605-e10605 (2024-01-09)
Primary T cell has been transformed into a cell-based delivery platform that synthesizes complex biologics at the disease site with spatiotemporal resolution. This broadly applicable technology can circumvent toxicities due to systemic administration of biologics that necessitates the use of
Mor Angel et al.
Nucleic acids research, 52(9), 5356-5375 (2024-02-17)
Stress granules (SGs) are cytoplasmic assemblies formed under various stress conditions as a consequence of translation arrest. SGs contain RNA-binding proteins, ribosomal subunits and messenger RNAs (mRNAs). It is well known that mRNAs contribute to SG formation; however, the connection between SG

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service