Skip to Content
Merck
All Photos(2)

Key Documents

526561

Sigma-Aldrich

PI 3-K/mTOR Inhibitor III, PKI-179

The PI 3-K/mTOR Inhibitor III, PKI-179, also referenced under CAS 1197160-28-3, controls the biological activity of PI 3-K/mTOR. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PI 3-K/mTOR Inhibitor III, PKI-179

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€213.00

€213.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€213.00

About This Item

Empirical Formula (Hill Notation):
C25H28N8O3 · xH2O
CAS Number:
Molecular Weight:
488.54 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€213.00


Estimated to ship on24 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=NC=C1)NC2=CC=C(C=C2)C3=NC(N4[C@H]5COC[C@@H]4CC5)=NC(N6CCOCC6)=N3.[xH2O]

InChI

1S/C25H28N8O3/c34-25(28-19-7-9-26-10-8-19)27-18-3-1-17(2-4-18)22-29-23(32-11-13-35-14-12-32)31-24(30-22)33-20-5-6-21(33)16-36-15-20/h1-4,7-10,20-21H,5-6,11-16H2,(H2,26,27,28,34)

InChI key

WXUUCRLKXQMWRY-UHFFFAOYSA-N

General description

A cell-permeable triazinyl-phenyl-pyridinylurea compound that acts as a potent inhibitor against mTOR (IC50 = 0.42 nM) and PI 3-K (IC50 = 8, 24, 74, 77, 14, and 11 nM against isoform α, β, γ, δ, α E545K mutant, and α H1047R mutant, respectively) activities, exhibiting much reduced or little activity against a panel of 361 other kinases (IC50 >50 µM), Cytochrome CYP isoforms, or hERG. It is reported to be orally available in mouse, rat, monkey, and dog species, and effectively suppress cancer growth both in cultures in vitro (IC50 = 22 and 29 nM, respectively, against MDA361 and PC3mm2) and in mice in vivo (by 30% of control MDA361 tumor size on day 42; 50 mg/kg/day p.o.) via apoptosis induction as a result of Akt/mTOR pathway inhibition.
The PI 3-K/mTOR Inhibitor III, PKI-179, also referenced under CAS 1197160-28-3, controls the biological activity of PI 3-K/mTOR. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Venkatesan, A.M., et al. 2010. Bioorg. Med. Chem. Lett.20, 5869.
Chen, Z., et al. 2010. J. Org. Chem.75, 1643.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service