Skip to Content
Merck
All Photos(1)

Key Documents

521233

Sigma-Aldrich

PDGFR Tyrosine Kinase Inhibitor IV

Synonym(s):

PDGFR Tyrosine Kinase Inhibitor IV, 3-Fluoro-N-(6,7-dimethoxy-2,4-dihydroindeno[1,2-c]pyrazol-3-yl)phenylamine, JNJ-10198409

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€200.00

€200.00


Estimated to ship on10 May 2025



Select a Size

Change View
1 MG
€200.00

About This Item

Empirical Formula (Hill Notation):
C18H16FN3O2
CAS Number:
Molecular Weight:
325.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€200.00


Estimated to ship on10 May 2025


Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mg/mL
methanol: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Fc1cc(ccc1)Nc2[nH]nc3c2Cc4c3cc(c(c4)OC)OC

InChI

1S/C18H16FN3O2/c1-23-15-7-10-6-14-17(13(10)9-16(15)24-2)21-22-18(14)20-12-5-3-4-11(19)8-12/h3-5,7-9H,6H2,1-2H3,(H2,20,21,22)

InChI key

ZDNURMVOKAERHZ-UHFFFAOYSA-N

General description

A cell-permeable indenopyrazole compound that displays potent antiproliferative properties in several human tumor cell lines (IC50<33 nM). Shown to act as an ATP-competitive and reversible inhibitor of PDGFR (IC50 = 4.2 nM and 45 nM for -β and -α, respectively) and c-Abl (IC50 = 22 nM). Exhibits less activity towards Lck, c-Src, and Fyn (IC50 = 100 nM, 185 nM and 378 nM, respectively) and exhibits little or no inhibition toward VEGFR, HER-2, Cdk′s-1,-2,-4 &-7, bFGFR-1 and EGFR (IC50 = 3.1 µM, >10 µM, >10 µM, 45.8 µM and >100 µM, respectively).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PDGFR Tyrosine Kinase
Product competes with ATP.
Reversible: yes
Target IC50: 4.2 nM and 45 nM for -β and -α form of PDGFR, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Ho, C.Y., et al. 2005. J. Med. Chem.48, 8163.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prem Swaroop Yadav et al.
iScience, 26(9), 107548-107548 (2023-08-28)
Low circulating phosphate (Pi) leads to rickets, characterized by expansion of the hypertrophic chondrocytes (HCs) in the growth plate due to impaired HC apoptosis. Studies in HCs demonstrate that Pi activates the Raf/MEK/ERK1/2 and mitochondrial apoptotic pathways. To determine how
Danielle Park et al.
Nature materials, 19(2), 227-238 (2019-10-30)
The isotropic or anisotropic organization of biological extracellular matrices has important consequences for tissue function. We study emergent anisotropy using fibroblasts that generate varying degrees of matrix alignment from uniform starting conditions. This reveals that the early migratory paths of
Vasiliki Tsata et al.
Developmental cell, 56(4), 509-524 (2021-01-08)
In mammals, perivascular cell-derived scarring after spinal cord injury impedes axonal regrowth. In contrast, the extracellular matrix (ECM) in the spinal lesion site of zebrafish is permissive and required for axon regeneration. However, the cellular mechanisms underlying this interspecies difference

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service