Skip to Content
Merck

Skip To

516705

o-Phenanthroline

≥99% (titration), Metal chelator, solid

Synonym(s):

o-Phenanthroline, 1,10-Phenanthroline Monohydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mg
Check Cart for Availability
€ 82,20

About This Item

Empirical Formula (Hill Notation):
C12H8N2 · xH2O
CAS Number:
Molecular Weight:
180.21 (anhydrous basis)
UNSPSC Code:
51111800
NACRES:
NA.77
MDL number:
Assay:
≥99% (titration)
Form:
solid
Storage condition:
OK to freeze, protect from light

€ 82,20


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

o-Phenanthroline, A metal chelating agent that prevents the induction of chromosomal aberrations in streptozotocin-treated cells.

Quality Segment

description

Merck USA index - 14, 7214

assay

≥99% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

white to cream

solubility

DMSO: 100 mg/mL, ethanol: 25 mg/mL, water: 3 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

n1c2c3ncccc3ccc2ccc1.O

InChI

1S/C12H8N2.H2O/c1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1;/h1-8H;1H2

InChI key

PPQJCISYYXZCAE-UHFFFAOYSA-N

General description

A metal chelating agent that prevents the induction of chromosomal aberrations in streptozotocin-treated cells. Acts as a general matrix metalloproteinase inhibitor. Forms a red chelate with Fe2+ that absorbs maximally at 510 nm. Can be used as a photometric reagent for iron.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Metalloprotease inhibitor
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Stix, B., et al. 2001. Am. J. Pathol.159, 561.
Bolzan, A.D., et al. 2000. Mutat. Res.447, 221.
Wang, Y., et al. 1999. J. Biol. Chem.274, 33043.
Harayama, T., et al. 1999. Jpn. J. Can. Res.90, 942.
Krane, S.M., et al. 1996. J. Biol. Chem.271, 28509.
Uhm, J.H., et al. 1996. Clin. Exp. Metastasis14, 421.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Toxic (F)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
164610234115512732
assay

≥99% (titration)

assay

≥90% (HPLC)

assay

≥94% (HPLC)

assay

≥97% (HPLC)

form

solid

form

solid

form

powder

form

solid

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

storage temp.

10-30°C

storage temp.

10-30°C

storage temp.

10-30°C

storage temp.

−20°C

storage condition

OK to freeze, protect from light

storage condition

OK to freeze

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, protect from light


pictograms

Skull and crossbonesEnvironment

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters