Skip to Content
Merck
All Photos(1)

Key Documents

5.09884

Sigma-Aldrich

Myristoylated-FEEERA, mP6

Synonym(s):

Myristoylated-FEEERA, mP6, Integrin Signaling Inhibitor I, mP6

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€230.00

€230.00


Estimated to ship on10 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€230.00

About This Item

Empirical Formula (Hill Notation):
C47H75N9O14
Molecular Weight:
990.15
UNSPSC Code:
12352200
NACRES:
NA.77

€230.00


Estimated to ship on10 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

−70°C

General description

A cell-permeable myristoylated hexapeptide derived from EXE-motif-containing integrin β3 subunit that selectively inhibits integrin outside-in signaling without affecting inside-out signaling . Shown to specifically block the interaction of Gα3 and integrin β3 subunit resulting in the inhibition of Gα13-dependent early phase outside-in signaling. Selectively inhibits thrombosis without affecting haemostasis.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
platelet integrin →IIb→3
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

FEEERA

Reconstitution

Following reconstitution, aliquot and freeze (-207deg;C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Shen, B., et al. 2013. Nature503, 131.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service