Skip to Content
Merck
All Photos(1)

Key Documents

5.09455

Sigma-Aldrich

Etomoxir

≥98% (HPLC), liquid, CPT-1 inhibitor, Calbiochem®

Synonym(s):

InSolution Etomoxir, R(+)-2-[6-(4-Chlorophenoxy)hexyl]-oxirane-2-carboxylic acid sodium salt hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
€195.00

€195.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
2 MG
€195.00

About This Item

Empirical Formula (Hill Notation):
C15H18ClO4 · Na
CAS Number:
Molecular Weight:
320.74
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€195.00


Estimated to ship on29 May 2025


Request a Bulk Order

Product Name

Etomoxir, InSolution, ≥98%, 50 mM aqueous solution, shown to inhibit carnitine palmitoyltransferase (CPT)-1 and DGAT activity

Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
protect from light

storage temp.

−70°C

SMILES string

[Na+].Clc1ccc(cc1)OCCCCCC[C@]2(OC2)C(=O)[O-]

InChI

1S/C15H19ClO4.Na/c16-12-5-7-13(8-6-12)19-10-4-2-1-3-9-15(11-20-15)14(17)18;/h5-8H,1-4,9-11H2,(H,17,18);/q;+1/p-1/t15-;/m1./s1

InChI key

RPACBEVZENYWOL-XFULWGLBSA-M

General description

A cell-permeable, irreversible, and stereospecific compound that is shown to inhibit carnitine palmitoyltransferase (CPT)-1 and DGAT activity in the mitochondria of rat heart H9c2 myoblastic cells at a concentration of 1-80 µM and 40 µM, respectively.

Biochem/physiol Actions

Primary Target
CPT1

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

A 50 mM (2 mg/118 µL) solution of Etomoxir (Cat. No. 236020) in H₂O.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C). Aliquots are stable for up to 6 months at -70°C.

Other Notes

Xu, F.Y., et al. 2003. J. Lipid Res.44, 415.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service