Skip to Content
Merck
All Photos(1)

Key Documents

5.08514

Sigma-Aldrich

Psalmotoxin-1

Synonym(s):

Psalmotoxin-1, ASIC1a Inhibitor, Psalmotoxin 1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€68.40

€68.40


Estimated to ship on25 May 2025



Select a Size

Change View
20 μG
€68.40

About This Item

Empirical Formula (Hill Notation):
C200H312N62O57S6
CAS Number:
Molecular Weight:
4689.39
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€68.40


Estimated to ship on25 May 2025


Assay

≥94% (HPLC)

Quality Level

form

semisolid
viscous liquid

potency

0.9 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

colorless

storage temp.

−20°C

General description

A potent proton-gated sodium channel (ASIC1a) channel blocking neurotoxin (IC50 = 0.9 nm) isolated from the Trinidad chevron tarantula. Psalmotoxin-1 (PcTx1) can distinguish between the two ASIC1 splice variants, ASIC1a and ASIC1b. Binding site is blocked when dimerization with ASIC2a or ASIC3.

Biochem/physiol Actions

Primary Target
ASIC1a

Warning

Toxicity: Standard Handling (A)

Sequence

EDCIPKWKGCVNRHGDCCEGLECWKRRRSFEVCVPKTPKT (disulfide bond 3-18, 10-23, 17-33)

Other Notes

Qadri, Y. J. et al. 2009. J. Biol. Chem.284, 17625.
M. Mazzuca, et al. 2007. Nat Neurosci.10, 943.
Escoubas, P. et al. 2000. J. Biol. Chem.275, 25116.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service