Skip to Content
Merck
All Photos(1)

Key Documents

5.06306

Sigma-Aldrich

IKKε/TBK1 Inhibitor II, MRT67307

Synonym(s):

IKKε/TBK1 Inhibitor II, MRT67307, N-(3-(5-Cyclopropyl-2-(3-(morpholinomethyl)phenylamino)pyrimidin-4-ylamino)propyl)cyclobutanecarboxamide, TANK Binding Kinase 1/IKKinducible Inhibitor II, TANK Binding Kinase 1/IKKinducible Inhibitor II, N-(3-(5-Cyclopropyl-2-(3-(morpholinomethyl)phenylamino)pyrimidin-4-ylamino)propyl)cyclobutanecarboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€214.00

€214.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€214.00

About This Item

Empirical Formula (Hill Notation):
C26H36N6O2
CAS Number:
Molecular Weight:
464.60
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

€214.00


Estimated to ship on28 May 2025


Request a Bulk Order

Assay

≥95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

amber

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

N5(CCOCC5)Cc1cc(ccc1)Nc2nc(c(cn2)C4CC4)NCCCNC(=O)C3CCC3

InChI

1S/C26H36N6O2/c33-25(21-5-2-6-21)28-11-3-10-27-24-23(20-8-9-20)17-29-26(31-24)30-22-7-1-4-19(16-22)18-32-12-14-34-15-13-32/h1,4,7,16-17,20-21H,2-3,5-6,8-15,18H2,(H,28,33)(H2,27,29,30,31)

InChI key

UKBGBACORPRCGG-UHFFFAOYSA-N

General description

A cell-permeable BX795 analog (>Cat. No. 204001) that acts as a potent, ATP-competitive, and reversible dual kinase inhibitor of TBK1/IKKε (IC50 = 19 and 160 nM, respectively) with excellent selectivity over IKKα and IKKβ (IC50 >10 µM). Interacts with the TBK1 kinase dimer interface and stabilizes the inactive DFG-out conformation. Also blocks the activity of MARK (microtubule- associated protein (MAP)-microtubule affinity regulating kinase) 1, 2, 3, and 4 (IC50 = 27, 52, 36, and 41 nM, respectively), SIK2 (IC50 = 67 nM) and Aurora B, JAK2, and MLK1,3 (> than 90% inhibition at 1 µM) in a 108-kinase panel. Increases TNF-α-stimulated NF-κB-dependent gene transcription in wild-type macrophages and enhances CREB-dependent gene transcription by promoting dephosphorylation of CREB-regulated transcription coactivator (CRTC3). Shown to aid TLR-stimulated production of anti-Inflammatory cytokines in macrophages while suppressing the secretion of pro-inflammatory cytokines. In response to pro-inflammatory stimuli, pre-treatment of BX795 robustly suppresses the activation of JNK and p38α, whereas MRT67307 does not exhibit such off-target effects.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water and salt.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
TBK1 kinase dimer interface
Reversible: yes
Secondary Target
IKKe, MARK, SIK

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Bruni, D., et al. 2013. J. Immunol.190, 2844.
Tu, D., et al. Cell Reports3, 747.
Larabi, A., et al. Cell Reports3, 734.
Clark, K., et al. 2012. Proc. Natl. Acad. Sci. USA109, 16986.
Clark, K., et al. 2011. Proc. Natl. Acad. Sci. USA108, 17093.
Clark, K., et al. 2011. Biochem. J.434, 93.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service