Skip to Content
Merck
All Photos(1)

Key Documents

5.06130

Sigma-Aldrich

Reserpine

Synonym(s):

Reserpine, VMAT2 Inhibitor, Reserpine, Vesicular monoamine transporter (VMAT) Antagonist, Reserpine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€243.00

€243.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
250 MG
€243.00

About This Item

Empirical Formula (Hill Notation):
C33H40N2O9
CAS Number:
Molecular Weight:
608.68
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€243.00


Estimated to ship on30 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mM

storage temp.

2-8°C

SMILES string

N21[C@H](C[C@H]5[C@@H](C2)C[C@H]([C@@H]([C@H]5C(=O)OC)OC)OC(=O)c6cc(c(c(c6)OC)OC)OC)c3[nH]c4c(c3CC1)ccc(c4)OC

InChI

1S/C33H40N2O9/c1-38-19-7-8-20-21-9-10-35-16-18-13-27(44-32(36)17-11-25(39-2)30(41-4)26(12-17)40-3)31(42-5)28(33(37)43-6)22(18)15-24(35)29(21)34-23(20)14-19/h7-8,11-12,14,18,22,24,27-28,31,34H,9-10,13,15-16H2,1-6H3/t18-,22+,24-,27-,28+,31+/m1/s1

InChI key

QEVHRUUCFGRFIF-MDEJGZGSSA-N

General description

Binds the vesicular monoamine transporter (VMAT2) and acts as an irreversible transport inhibitor of biogenic amines. Used as a long-lasting bioamine depleter. Prevents bioamine transport into synaptic vesicles and chromaffin granules. Used as antihypertensive and antipsychotic drugs.

Biochem/physiol Actions

Primary Target
VMAT2

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Oe, T., et al. 2010. Neurosci.169, 1860.
Metzger, R. et al. 2002. Eur. J. Pharmacol.456, 39.
Heslop, E., et al. 1999. Neuropharmacol.38, 883.
Schuldiner, S., et al. 1993. J. Biol. Chem.268, 29.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Repr. 1A - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service