Skip to Content
Merck
All Photos(1)

Key Documents

5.06074

Sigma-Aldrich

PP2A Activating Ligand, ITH12246

Synonym(s):

PP2A Activating Ligand, ITH12246

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€195.00

€195.00


Estimated to ship on03 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€195.00

About This Item

Empirical Formula (Hill Notation):
C16H19N3O2 · xHCl
CAS Number:
Molecular Weight:
285.34 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

€195.00


Estimated to ship on03 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable, relatively non-toxic, 1,8-naphthyridine derived compound that blocks the inhibitory effect of protein phosphatase inhibitors, such as okadaic acid, on protein phosphatase 2A (PP2A). Exhibits blood-brain barrier permeability. Protects neurons against β-amyloid peptides (Ab1-42) toxicity and okadaic acid (Cat. No. 459620)-induced tau hyperphosphorylation. Also protects against rotenone and oligomycin A induced neurotoxicity in SH-SY5Y neuroblastoma cells (at ~300 nM). Prevents the development of glutamate-induced neuronal lesions in rat hippocampal slices by up-regulating PP2A (~ 3.0 µM). Shown to reverse the scopolamine-induced memory loss in mice (~10 mg/kg i.p) and significantly reduces the infarct volume in an animal model of stroke (~2.5 mg/kg). Also acts as an inhibitor of acetylcholinesterase activity in Electrophorus electricus (IC50 = 60 nM) and human erythrocytes (IC50 = 780 nM).

Biochem/physiol Actions

Primary Target
PP2A

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lorrio, S., et al. 2013. ACS Chem. Neurosci.4,1267.
de Los Rios, C., et al. 2010. J. Med. Chem.53,5129.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service