Skip to Content
Merck
All Photos(1)

Key Documents

5.05989

Sigma-Aldrich

Vigabatrin

Synonym(s):

Vigabatrin, 4-Amino-5-hexenoic acid, GABA Transaminase Inhibitor, Vigabatrin, γ-Vinyl-GABA, Sabril

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€101.00

€101.00


Estimated to ship on13 April 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€101.00

About This Item

Empirical Formula (Hill Notation):
C6H11NO2
CAS Number:
Molecular Weight:
129.16
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€101.00


Estimated to ship on13 April 2025


Request a Bulk Order

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

H2O: 100 mM

storage temp.

2-8°C

SMILES string

[N+H3]C(CCC(=O)[O-])C=C

InChI

1S/C6H11NO2/c1-2-5(7)3-4-6(8)9/h2,5H,1,3-4,7H2,(H,8,9)

InChI key

PJDFLNIOAUIZSL-UHFFFAOYSA-N

General description

A selective, irreversible inhibitor of GABA transaminase with antiepileptic properties. Covalently binds to the active site of GABA tansaminase to inactivate it in a dose-dependent manner and increases extracellular GABA levels in the brain. Rapidly absorbed following oral administration and is shown to inhibit [3H]GABA uptake across the apical membrane of Caco-2 cell monolayers in a concentration-dependent manner (IC50 = 2.9 mM)

Biochem/physiol Actions

Primary Target
GABA Transaminase
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Abbot, E.L., et al. 2006. Br. J. Pharmacol.147, 298.
Schmid, L., et al. 1996. J. Pharmacol. Exp. Therap.276, 977.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service