Skip to Content
Merck
All Photos(1)

Key Documents

5.05981

Sigma-Aldrich

PK11195

Synonym(s):

PK11195, TSPO Activator II, Mitochondrial Translocator Protein Activator II, PK 11195, PK-11195, PK11195

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€103.00

€103.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€103.00

About This Item

Empirical Formula (Hill Notation):
C21H21ClN2O
CAS Number:
Molecular Weight:
352.86
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€103.00


Estimated to ship on28 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

20 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mM

storage temp.

2-8°C

SMILES string

Clc1c(cccc1)c2nc(cc3c2cccc3)C(=O)N(C(CC)C)C

InChI

1S/C21H21ClN2O/c1-4-14(2)24(3)21(25)19-13-15-9-5-6-10-16(15)20(23-19)17-11-7-8-12-18(17)22/h5-14H,4H2,1-3H3

InChI key

RAVIZVQZGXBOQO-UHFFFAOYSA-N

General description

An isoquinoline carboxamide that acts as a potent ligand for peripheral bezodiazepine receptor (Kd = 20 nM). Exhibits anti-inflammatory properties. Binds to monocytes and following lesioning its binding up-regulated in brain macrophages and activated microglia. Shown to enhance the susceptibility of cells to apoptosis induction by DNA damaging agents and can reverse the suppression of apoptosis by Bcl-2. Serves as a ligand of the outer membrane translocator protein and reduces mitochondrial transmembrane potential within 30 minutes of exposure (˜100 µM).

Biochem/physiol Actions

Primary Target
peripheral bezodiazepine receptor

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Naoi, M., et al., 2013. J. Neural Transm.In press
Casellas, P., et al., 2002. Neurochem. Int.40, 475.

Waterfield, J.D., et al., 1999. Rheumtology38, 1068.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service