Skip to Content
Merck
All Photos(1)

Key Documents

5.05759

Sigma-Aldrich

MitoBloCK-6

Synonym(s):

MitoBloCK-6, 2-(((4-Anilinophenyl)imino)methyl)-4,6-dichlorophenol, 2,4-Dichloro-6-(((((phenylamino)phenyl)imino)methyl)phenol), Mitochondrial Protein Import Blocker from the Carla Koehler laboratory, MB6, MB-6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14Cl2N2O
CAS Number:
Molecular Weight:
357.23
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable, stable dichlorosalicylaldehyde Schiff′s base that acts as a potent, selective inhibitor of Mia40/Erv1 redox-mediated import pathway (IC50 = 700 nM, 900 nM, and 1.4 µM for ALR, Erv1, and Erv2, respectively). Significantly reduces the import of CX9 proteins, Erv1, Tim23, and ADP/ATP carrier (AAC). However, it does not affect mitochondrial membrane integrity as evidenced by the lack of aconitase, AAC, Tim54, Mia40, and cytochrome c release. Has no effect on protein disulfide isomerase, flavin adenine dinucleotide, and succinate dehydrogenase activities and does not disrupt mitochondrial net work or reduce viability of cells even at high concentrations (~100 µM in HeLa and HEK293 cells). Reported to specifically cause cytochrome c release, activate caspase-3, and induce apoptosis in human embryonic stem cells (~20 µM), but not in differentiated cells. Reversibly impairs cardiac development and reduces heart rate in zebra fish that is attributed to mitochondrial dysfunction.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Erv1
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Dabir, D.V., et al. 2013. Dev. Cell25, 81.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deepa V Dabir et al.
Developmental cell, 25(1), 81-92 (2013-04-20)
The mitochondrial disulfide relay system of Mia40 and Erv1/ALR facilitates import of the small translocase of the inner membrane (Tim) proteins and cysteine-rich proteins. A chemical screen identified small molecules that inhibit Erv1 oxidase activity, thereby facilitating dissection of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service