Skip to Content
Merck
All Photos(1)

Key Documents

5.05316

Sigma-Aldrich

PHA-543,613

Synonym(s):

PHA-543,613, N-[(3R)-1-Azabicyclo[2.2.2]oct-3-yl]furo[2,3-c]pyridine-5-carboxamide, Alpha 7 Nicotinic Acetylcholine Receptor (α7 nAChR) Agonist, PHA-543613

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17N3O2
CAS Number:
Molecular Weight:
271.31
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 20 mg/mL

storage temp.

2-8°C

InChI

1S/C15H17N3O2/c19-15(12-7-11-3-6-20-14(11)8-16-12)17-13-9-18-4-1-10(13)2-5-18/h3,6-8,10,13H,1-2,4-5,9H2,(H,17,19)/t13-/m0/s1

InChI key

IPKZCLGGYKRDES-ZDUSSCGKSA-N

General description

A potent agonist highly selective for α7 nicotinic acetylcholine receptor (α7 nAChR). Bioavaible admitted orally. Widely used in studies of the functional roles of α7 nAChR in learning and memory, sensory gating, and cognitive functions as well as Schizophrenia and Alzheimer′s disease.

Biochem/physiol Actions

Primary Target
α7 nAChR

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Mazurov, A., et al., 2011. J. Med. Chem.54, 7943.
Wishka, G., et al., 2006. J. Med. Chem.49, 4425.
Faghih, R., et al., 2008. J. Med. Chem.51, 701.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service