Skip to Content
Merck
All Photos(1)

Key Documents

5.05226

Sigma-Aldrich

Pancuronium Dibromide

Synonym(s):

Pancuronium Dibromide, 1,1ʹ-(3α,17β-Dihydroxy-2α,5β-androstan-2-β,16β-ylene) bis[1-methylpiperidinium] diacetate dibromide, Nicotinic Acetylchorine Receptor Antagonist, Pancuronium Dibromide, nAChR Antagonist, Pancuronium Dibromide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€68.90

€68.90


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€68.90

About This Item

Empirical Formula (Hill Notation):
C35H60N2O4 · Br2
CAS Number:
Molecular Weight:
732.67
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€68.90


Estimated to ship on28 May 2025


Request a Bulk Order

Assay

≥98% (NMR)

Quality Level

form

solid

potency

5.5 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

water: 100 mM

storage temp.

2-8°C

SMILES string

[Br-].[Br-].[N+]6(CCCCC6)([C@@H]1[C@@H]([C@@]2([C@H]([C@H]3[C@@H]([C@@]4([C@H](C[C@@H]([C@H](C4)[N+]5(CCCCC5)C)OC(=O)C)CC3)C)CC2)C1)C)OC(=O)C)C

InChI

1S/C35H60N2O4.2BrH/c1-24(38)40-32-21-26-13-14-27-28(35(26,4)23-31(32)37(6)19-11-8-12-20-37)15-16-34(3)29(27)22-30(33(34)41-25(2)39)36(5)17-9-7-10-18-36;;/h26-33H,7-23H2,1-6H3;2*1H/q+2;;/p-2/t26-,27+,28-,29-,30-,31-,32-,33-,34-,35-;;/m0../s1

InChI key

NPIJXCQZLFKBMV-YTGGZNJNSA-L

General description

A highly potent competitive antagonist selective for nAChRs (IC50 = 5.5 nM). A very potent non-depolarizing skeletal muscle relaxant but no sedative or analgesic effects.

Biochem/physiol Actions

Primary Target
nAChR

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lowenick et al., 2001. Eur. J. Pharmacol.413, 31.

Melnikov, L., et al., 1999. Gen Pharmacol.33, 313.

Maestrone, E., et al., 1994. Br. J. Pharmacol.111, 283.

Fryer, D., et al., 1987. Naunyn Schmiedebergs Arch. Pharmacol.335, 367.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service