Skip to Content
Merck
All Photos(1)

Key Documents

5.05225

Sigma-Aldrich

ISO-1

InSolution, ≥95%, 50 mM in DMSO, MIF Antagonist

Synonym(s):

InSolution MIF Antagonist, ISO-1, ( S, R)-3-(4-Hydroxyphenyl)-4,5-dihydro-5-isoxazole acetic acid, methyl ester, Methyl 2-(3-(4-hydroxyphenyl)-4,5-dihydroisoxazol-5-yl)acetate, Macrophage Migration Inhibitory Factor Antagonist, ISO-1, (S,R)-3-(4-Hydroxyphenyl)-4,5-dihydro-5-isoxazole acetic acid, methyl ester, Methyl 2-(3-(4-hydroxyphenyl)-4,5-dihydroisoxazol-5-yl)acetate, Macrophage Migration Inhibitory Factor Antagonist, ISO-1

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€203.00

€203.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€203.00

About This Item

Empirical Formula (Hill Notation):
C12H13NO4
CAS Number:
Molecular Weight:
235.24
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

€203.00


Estimated to ship on19 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

InChI

1S/C12H13NO4/c1-16-12(15)7-10-6-11(13-17-10)8-2-4-9(14)5-3-8/h2-5,10,14H,6-7H2,1H3

InChI key

AIXMJTYHQHQJLU-UHFFFAOYSA-N

General description

A cell-permeable isoxazoline compound that displays anti-inflammatory properties. Inhibits MIF dopachrome tautomerase activity by binding to its catalytic active site (IC50 = 7 µM for D-dopachrome tautomerase) and suppresses the production of TNFα, PGE2 and COX-2 in human monocytes and arachidonic acid in RAW 264.7 macrophages. Shown to exhibit antidiabetogenic properties in immunoinflammatory diabetic mouse model.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MIF dopachrome tautomerase

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 50 mM (10 mg/850 µL) sterile-filtered solution of MIF Antagonist, ISO-1 (Cat. No. 475837) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C). Aliquots are stable for up to 1 year at -70°C.

Other Notes

Cvetkovic, I., et al. 2005. Endocrinology146, 2942.
Lubetsky, J.B., et al. 2002. J. Biol. Chem.277, 24976.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service