5.04843
Fbxo3 Inhibitor, BC-1215
Synonym(s):
Fbxo3 Inhibitor, BC-1215, N¹,N²- bis(4-(Pyridin-2-yl)benzyl)ethane-1,2-diamine, F Box Only Protein 3 Inhibitor, F Box Protein 3 Inhibitor, BC1215, F Box and Leucine-rich Repeat Protein 2 Activator, Fbxl2 Activator
About This Item
Assay
≥98% (HPLC)
Quality Level
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
beige
solubility
DMSO: 10 mg/mL
storage temp.
−20°C
SMILES string
N(CCNCc3ccc(cc3)c4ncccc4)Cc1ccc(cc1)c2ncccc2
InChI key
IXEPQJQQSLMESJ-UHFFFAOYSA-N
General description
Biochem/physiol Actions
Fbox3
Packaging
Warning
Reconstitution
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service