Skip to Content
Merck
All Photos(1)

Key Documents

5.00653

Sigma-Aldrich

Nef Hck Activation Inhibitor, B9

Synonym(s):

Nef Hck Activation Inhibitor, B9, Nef Dimerization Blocker, ( E)-4-((3-Chlorophenyl)diazenyl)-5-hydroxy-3-(4-nitrophenyl)-1H-pyrazole-1-carbothioamide, Nef Dimerization Blocker, (E)-4-((3-Chlorophenyl)diazenyl)-5-hydroxy-3-(4-nitrophenyl)-1H-pyrazole-1-carbothioamide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€170.00

€170.00


Estimated to ship on27 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€170.00

About This Item

Empirical Formula (Hill Notation):
C16H11ClN6O3S
CAS Number:
Molecular Weight:
402.81
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

€170.00


Estimated to ship on27 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 2.5 mg/mL

storage temp.

2-8°C

SMILES string

C1=CC(=CC(=C1)Cl)N=NC2=C(NN(C2=O)C(=S)N)C3=CC=C(C=C3)[N+](=O)[O-]

InChI

1S/C16H11ClN6O3S/c17-10-2-1-3-11(8-10)19-20-14-13(21-22(15(14)24)16(18)27)9-4-6-12(7-5-9)23(25)26/h1-8,21H,(H2,18,27)

InChI key

LEQSBTDRMOZWRP-UHFFFAOYSA-N

General description

A cell-permeable diphenylpyrazolo compound that is shown to disrupt HIV accessary protein Nef dimerization (IC50 = 3 µM in HEK293T cells) and prevent Nef-mediated Src family kinase (SFK) Hck activation (IC50 = 2.8 µM) without directly affecting the catalytic activity of c-Src, Hck, Lck, or Lyn (IC50 >20 µM). Shown to effectively suppress HIV and SIV viral replication (IC50<0.3 µM in 9 d HIV-1 NL4-3-infected CEM-T4 cultures; IC50 = 1 µM in 5 d SIV ΔB670-infected CEM-174 cultures) and infectivity (45% and 50% inhibition against HIV-1 NL4-3 and SIV ΔB670, respectively, following 48 h 3 µM B9 treatment in TZM-bl reporter cells) in a Nef-dependent manner, being ineffective against the replication of Nef-defective HIV-1 mutant. Molecular docking studies predict a high-affinity B9-targeting site formed at the Nef dimerization interface and a low-affinity B9-binding site on each Nef monomer at the dimer interface periphery, with Asn126 contributing to both modes of binding. Consistently, in vitro binding studies reveal two B9 KD values of 1.72 nM and 860 nM toward full-length HIV-1 Nef and a complete loss of B9 binding toward Nef N126A/L/Q mutants.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Nef
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Emert-Sedlak, L.A., et al. 2013. Chem. Biol.20, 82.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service