Skip to Content
Merck
All Photos(1)

Key Documents

5.00649

Sigma-Aldrich

HMTase Inhibitor IX, MM-102

Synonym(s):

HMTase Inhibitor IX, MM-102, Histone Methyltransferase Inhibitor IX, MM102, ( S)-N-( bis(4-Fluorophenyl)methyl)-1-(2-(2-ethyl-2-isobutyramidobutanamido)-5-guanidinopentanamido)cyclopentanecarboxamide, TFA, Histone Methyltransferase Inhibitor IX, MM102, (S)-N-(bis(4-Fluorophenyl)methyl)-1-(2-(2-ethyl-2-isobutyramidobutanamido)-5-guanidinopentanamido)cyclopentanecarboxamide, TFA

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€244.00

€244.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€244.00

About This Item

Empirical Formula (Hill Notation):
C35H49F2N7O4 · xC2HF3O2
Molecular Weight:
669.80 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

€244.00


Estimated to ship on25 May 2025


Request a Bulk Order

Assay

≥95% (HPLC)

Quality Level

form

powder

potency

1 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

light beige

solubility

DMSO: 100 mg/mL

storage temp.

−20°C

General description

A cell-permeable peptidomimetic that competes against MLL1 (Mixed Lineage Leukemia 1) minimum essential WIN (WDR5 INteracting) motif (-Ala3764-Arg-Ala3766-) for WDR5 (Trp-Asp Repeat Domain 5) binding (IC50/Ki = 2.5 nM/<1 nM and 540 nM/120 nM, respectively), effectively preventing MLL1 from complex assembling with WDR5, RbBP5 (Retinoblastoma Binding Protein 5), and ASH2L (Absent Small or Homeotic-2-Like) for enhanced H3K4 methyltransferase activity (IC50 = 0.4 nM). Shown to reduce HoxA9 and Meis-1 mRNA expression (by 76% and 36%, respectively, after 96 h 50 µM MM-102 treatment) in myeloblasts derived from MLL1-AF9-transduced murine bone marrow cells and preferentially inhibit the growth of MLL1-AF4-harboring MV4;11 and MLL1-ENL-harboring KOPN8 leukemia cultures (GI50 in 7 days = 25 µM) via apoptosis induction (by >75% in 4 days in MV4:11 cultures; 50 µM MM-102), while exhibiting much reduced potency against BCR-ABL-dependent K562 cells (GI50 in 7 days = 84 µM; >90 live cells after 4-day 50 µM MM-102 treatment).
A cell-permeable peptidomimetic that prevents MLL1 (Mixed Lineage Leukemia 1) from complex assembling with WDR5 (Trp-Asp Repeat Domain 5), RbBP5 (Retinoblastoma Binding Protein 5), and ASH2L (Absent Small or Homeotic-2-Like) for enhanced H3K4 methyltransferase activity (IC50 = 0.4 nM). Shown to reduce HoxA9 and Meis-1 mRNA expression in myeloblasts derived from MLL1-AF9-transduced murine bone marrow cells (50 µM for 96 h) and preferentially inhibit the growth of MLL1-AF4-harboring MV4;11 and MLL1-ENL-harboring KOPN8 leukemia cultures (GI50 in 7 days = 25 µM) via apoptosis induction, while exhibiting much reduced potency against BCR-ABL-dependent K562 cells (GI50 in 7 days = 84 µM).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
WDR5
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Karatas, H., et al. 2013. J. Am. Chem. Soc.135, 669.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service