Skip to Content
Merck
All Photos(1)

Key Documents

5.00500

Sigma-Aldrich

MALT1 Inhibitor II, Mepazine Hydrochloride

Synonym(s):

MALT1 Inhibitor II, Mepazine Hydrochloride, 10-((1-Methylpiperidin-3-yl)methyl)-10H-phenothiazine, Hydrochloride, 10-((1-Methyl-3-piperidyl)methyl)phenothiazine, Hydrochloride, 10-((1-Methyl-3-piperidinyl)methyl)-10H-phenothiazine, Hydrochloride, MALT-1 Inhibitor II, Mucosa-Associated Lymphoid tis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23ClN2S
CAS Number:
Molecular Weight:
346.92
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

gray to off-white

solubility

H2O: 2.5 mg/mL
DMSO: 25 mg/mL

storage temp.

−20°C

InChI

1S/C19H22N2S.Cl/c1-20-12-6-7-15(13-20)14-21-16-8-2-4-10-18(16)22-19-11-5-3-9-17(19)21;/h2-5,8-11,15H,6-7,12-14H2,1H3;

InChI key

ZGQUIYCUOPQIDJ-UHFFFAOYSA-N

General description

Mepazine hydrochloride is a cell-permeable phenothiazine derivative that inhibits mucosa-associated lymphoid tis 1(MALT1). It does not inhibit the caspase 3 or 8 proteolytic activity in a reversible and non-competitive manner. It blocks anti-CD3/CD28-stimulated IL-6 production in primary murine CD4+ T-cell and human peripheral blood mononuclear cell (PBMC) cultures. Mepazine has been observed to inhibit the MALT1 activity in activated B cell-like diffuse large B cell lymphoma (ABC-DLBCL)and RelB cleavage, resulting in effective blockage of nuclear factor (NF-κB)-dependent interleukin-6, 10 as well as anti-apoptoticB-cell lymphoma - extra-large (Bcl-xL) and FLIP-L expression. Mepazineshows ABC-DLBCL-selective cytotoxicity over GCB-DLBCL both in cultures in vitro and in mice in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MALT1

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as a hydrochloride salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

The molecular weight for this compound isbatch-specific due to variable water content. Please refer to the vial label orthe certificate of analysis for the batch-specific molecular weight. Themolecular weight provided represents the baseline molecular weight withoutwater.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leonie Konczalla et al.
International journal of cancer, 146(6), 1618-1630 (2019-07-11)
MALT1 is a key mediator of NF-κB signaling and a main driver of B-cell lymphomas. Remarkably, MALT1 is expressed in the majority of pancreatic ductal adenocarcinomas (PDACs) as well, but absent from normal exocrine pancreatic tissue. Following, MALT1 shows off
Conor Mc Guire et al.
Journal of neuroinflammation, 11, 124-124 (2014-07-22)
The paracaspase mucosa-associated lymphoid tissue lymphoma translocation protein 1 (MALT1) is crucial for lymphocyte activation through signaling to the transcription factor NF-κB. Besides functioning as a scaffold signaling protein, MALT1 also acts as a cysteine protease that specifically cleaves a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service