Skip to Content
Merck
All Photos(1)

Key Documents

4954

Sigma-Aldrich

Oleic Acid

Synonym(s):

Oleic Acid, cis-9-Octadecenoic Acid, 18:1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H34O2
CAS Number:
Molecular Weight:
282.46
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

Assay

≥99% (GC)

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

colorless

solubility

chloroform: 10 mg/mL
ethanol: 5 mg/mL

density

0.89 g/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

OC(=O)CCCCCCC\C=C\CCCCCCCC

InChI

1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H,19,20)/b10-9+

InChI key

ZQPPMHVWECSIRJ-MDZDMXLPSA-N

General description

A cis-unsaturated fatty acid that has been shown to activate protein kinase C in hepatocytes. Potentiates acetylcholine receptor currents by activating CaM kinase II, independent of the PKC pathway.
Unsaturated fatty acid that has been shown to activate protein kinase C in hepatocytes. Density: 0.89 g/ml.

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Nishizaki, T., et al. 1997. NeuroReport 8, 597.
Bessesen, D.H., et al. 1993. J. Lipid Res.34, 229.
Felden, F., et al. 1993. Biochem. Biophys. Res. Commun.190, 602.
Williams, L.L., et al. 1993. Proc. Soc. Exp. Biol. Med.202, 239.
Diaz-Guerra, M.J. 1991. J. Biol. Chem.266, 23568.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Felden et al.
Biochemical and biophysical research communications, 190(2), 602-608 (1993-01-29)
The free fatty acid (FFA) concentration in the epididymal cytosol of the adult rat was found to be 20-fold higher than in the serum. The binding of [3H] dihydrotestosterone to epididymal rat androgen binding protein (rABP) was modified by physiological
M J Díaz-Guerra et al.
The Journal of biological chemistry, 266(35), 23568-23576 (1991-12-15)
The effect of oleate on the subcellular distribution of protein kinase C (PKC) was studied in isolated hepatocytes and in perfused rat liver in the presence of physiological concentrations of serum albumin. A time- and dose-dependent translocation of PKC from
L L Williams et al.
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.), 202(2), 239-245 (1993-02-01)
The polyunsaturated omega-6 fatty acid, arachidonic acid ([AA] 20:4n-6), is both the key of the immunoregulatory substances, prostaglandins, and leukotrienes, and an essential component of immune cell membrane phospholipids, providing stability and flexibility to ensure cellular function. To explore possible
T Nishizaki et al.
Neuroreport, 8(3), 597-601 (1997-02-10)
Oleic acid, a cis-unsaturated free fatty acid, is proposed to be involved in the protein kinase C (PKC) activation pathway. Its biological actions, however, have not been well-characterized. We examined the effects of oleic acid on acetylcholine (ACh)-gated channel currents
D H Bessesen et al.
Journal of lipid research, 34(2), 229-238 (1993-02-01)
Lipoprotein lipase (LPL) is important for the delivery of triglyceride fatty acids (TGFA) to a variety of tissues. We have used measurements of heparin-releasable LPL activity, immunohistochemistry, in situ hybridization, and Northern analysis to more fully characterize the location of

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service