Skip to Content
Merck
All Photos(1)

Key Documents

4.90189

Sigma-Aldrich

1-Ethyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide

for synthesis

Synonym(s):

1-Ethyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11F6N3O4S2
CAS Number:
Molecular Weight:
391.31
MDL number:
UNSPSC Code:
12352107

Quality Level

Assay

≥98.0% (HPLC)

form

liquid

potency

50-300 mg/kg LD50, oral (Rat)

mp

-15 °C

density

1.52 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C6H11N2.C2F6NO4S2/c1-3-8-5-4-7(2)6-8;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h4-6H,3H2,1-2H3;/q+1;-1

InChI key

LRESCJAINPKJTO-UHFFFAOYSA-N

Analysis Note

Purity (HPLC): ≥ 98.0 %
Identity (NMR): passes test
Halides (IC): ≤ 0.1 %
Water (K. F.): ≤ 1.0 %

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service