Skip to Content
Merck
All Photos(1)

Key Documents

4.90054

Sigma-Aldrich

1-Ethyl-3-methylimidazolium chloride

for synthesis

Synonym(s):

1-Ethyl-3-methylimidazolium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11ClN2
CAS Number:
Molecular Weight:
146.62
MDL number:
UNSPSC Code:
12352107
Pricing and availability is not currently available.

Assay

≥98.0% (HPLC)

mp

77-79 °C

transition temp

flash point 186 °C

storage temp.

2-30°C

InChI

1S/C6H11N2/c1-3-8-5-4-7(2)6-8/h4-6H,3H2,1-2H3/q+1

InChI key

NJMWOUFKYKNWDW-UHFFFAOYSA-N

Analysis Note

Purity (HPLC): ≥ 98.0 %
Identity (NMR): passes test
Water (K. F.): ≤ 1.0 %

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

370.4 °F - closed cup

Flash Point(C)

188 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service