Skip to Content
Merck
All Photos(1)

Key Documents

481984

Sigma-Aldrich

Nifuroxazide

A cell-permeable and orally available nitrofuran-based antidiarrheal agent that effectively suppresses the activation of cellular STAT1/3/5 transcription activity.

Synonym(s):

Nifuroxazide, 5-Nitro-2-furaldehyde- p-hydroxybenzoylhydrazone, 5-Nitro-2-furaldehyde-p-hydroxybenzoylhydrazone

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€87.00

€87.00


Estimated to ship on01 June 2025



Select a Size

Change View
500 MG
€87.00

About This Item

Empirical Formula (Hill Notation):
C12H9N3O5
CAS Number:
Molecular Weight:
275.22
MDL number:
UNSPSC Code:
12352125
NACRES:
NA.77

€87.00


Estimated to ship on01 June 2025


Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

bright yellow

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[N+](=O)([O-])c1[o]c(cc1)\C=N\NC(=O)c2ccc(cc2)O

InChI

1S/C12H9N3O5/c16-9-3-1-8(2-4-9)12(17)14-13-7-10-5-6-11(20-10)15(18)19/h1-7,16H,(H,14,17)/b13-7+

InChI key

YCWSUKQGVSGXJO-NTUHNPAUSA-N

General description

A cell-permeable and orally available nitrofuran-based antidiarrheal agent that effectively suppresses the activation of cellular STAT1/3/5 transcription activity (IC50 = 3 µM against IL-6-induced STAT3 activation in U3A cells), while exhibiting little effect against TNF-α-induced NF-κB activation in 293 cells. Shown to inhibit cellular tyrosine phosphorylations of JAK2/TYK2/STAT3 (IC50 = 10 µM against tyr705 of STAT3 in U266 cells), but not those of Akt/EGFR/JAK1/MAPK/Src or the ser727 phosphorylation of STAT3.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Nelson, E.A., et al. 2008. Blood112, 5095.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sunyoung Seo et al.
Biomedicines, 10(11) (2022-11-12)
Cancer stem-like cells (CSCs) are considered promising targets for anti-cancer therapy owing to their role in tumor progression. Extensive research is, therefore, being carried out on CSCs to identify potential targets for anti-cancer therapy. However, this requires the availability of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service