Skip to Content
Merck
All Photos(1)

Key Documents

481915

Millipore

NAD+, Lithium Salt

A major electron acceptor molecule in biological oxidations. Spectra (pH 7.0): 250/260 nm: 0.76-0.86; 280/260 nm: 0.18-0.28.

Synonym(s):

NAD+, Lithium Salt, β-Nicotinamide-adenine Dinucleotide, Li

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
Inquire


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
1 G
Inquire

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.51


Estimated to ship on02 June 2025


Request a Bulk Order

Quality Level

description

Merck USA index - 14, 6344

Assay

≥90% (enzymatic)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

[Li+].[P](=O)(O[P](=O)(OC[C@H]3O[C@H]([C@@H]([C@@H]3O)O)[n]4c5ncnc(c5nc4)N)O)(OC[C@H]1O[C@H]([C@@H]([C@@H]1[O-])[O-])[n+]2cc(ccc2)C(=O)N)O

InChI

1S/C21H26N7O14P2.Li/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33;/h1-4,7-8,10-11,13-16,20-21,30,32H,5-6H2,(H2,23,33)(H,34,35)(H,36,37)(H2,22,24,25);/q-1;+1/t10-,11-,13-,14-,15-,16-,20-,21-;/m1./s1

InChI key

VKCIQQTYIUNWEW-QYZPTAICSA-N

General description

A major electron acceptor molecule in biological oxidations (spectra: 0.76-0.86 at 250/260 nm, pH 7.0; 0.18-0.28 at 280/260 nm, pH 7.0).
Spectra (pH 7.0): 250/260 nm: 0.76-0.86; 280/260 nm: 0.18-0.28.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Avoid freeze/thaw cycles of solutions.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service