Skip to Content
Merck

Skip To

4.80855

Citric acid 50%

EMPROVE® EXPERT

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 L
Check Cart for Availability
€ 55,10
25 L
180 L
950 L
Check Cart for Availability
€ 15.970,00

About This Item

UNSPSC Code:
12352106

€ 55,10


Check Cart for Availability

Request a Bulk OrderRequest a Sample
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

EMPROVE® EXPERT

assay

50%

form

liquid

density

1.24 g/cm3 at 20 °C

application(s)

cleaning in place
pharma/biopharma processes

storage temp.

15-25°C

General description

Every bio-manufacturing process involves cleaning to protect equipment and prevent cross-contamination. All our GMP (IPEC/EXCiPACT) solutions for CIP and buffer preparation are manufactured under Excipient GMP conditions and feature low bioburden levels, backed by our Emprove® Program for exceptional quality and documentation support.
As a result, you can integrate our solutions directly into your biopharmaceutical production process. In addition to our comprehensive portfolio, we also offer custom manufacturing services to meet your exact needs.

Application

Citric acid 50% is used in the CIP process of biopharma manufacturing to effectively lower pH, chelate metal ions, and dissolve mineral deposits, ensuring thorough cleaning of equipment.

Features and Benefits

  • Comprehensive, up-to-date documentation you need to navigate regulatory challenges, manage risks, and improve processes.
  • Suitable for higher-risk applications where the lowest microbiological and endotoxin levels are of utmost importance.
  • Emprove® products are designed to facilitate compliance with global regulations (e.g., ICH Q7, EU GMP, USP, EP).
  • Detailed supply chain and manufacturing information helps ensure traceability and continuity, which is critical for GMP manufacturing.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.151871.0023827487
form

liquid

form

liquid

form

liquid

form

powder or crystals

product line

EMPROVE® EXPERT

product line

-

product line

EMPROVE® EXPERT

product line

BioUltra

density

1.24 g/cm3 at 20 °C

density

1.41 g/cm3 at 20 °C

density

1.52 g/cm3 at 20 °C

density

-

storage temp.

15-25°C

storage temp.

2-30°C

storage temp.

15-25°C

storage temp.

-

Quality Level

500

Quality Level

100

Quality Level

500

Quality Level

200


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

12 - Non Combustible Liquids

hcodes



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters