Skip to Content
Merck
All Photos(1)

Key Documents

475990

Sigma-Aldrich

mTOR Inhibitor IV, Ku-63794

The mTOR Inhibitor IV, Ku-63794, also referenced under CAS 938440-64-3, controls the biological activity of mTOR. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

mTOR Inhibitor IV, Ku-63794, rel-5-[2-[(2R,6S)-2,6-Dimethyl-4-morpholinyl]-4-(4-morpholinyl)pyrido[2,3-d]pyrimidin-7-yl]-2-methoxybenzenemethanol, Ku-0063794, mTOR Inhibitor IV, Ku-63794

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€188.00

€188.00


Estimated to ship on16 May 2025



Select a Size

Change View
5 MG
€188.00

About This Item

Empirical Formula (Hill Notation):
C25H31N5O4
CAS Number:
Molecular Weight:
465.54
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€188.00


Estimated to ship on16 May 2025


Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 15 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N5(C[C@H](O[C@H](C5)C)C)c1nc2nc(ccc2c(n1)N4CCOCC4)c3cc(c(cc3)OC)CO

InChI

1S/C25H31N5O4/c1-16-13-30(14-17(2)34-16)25-27-23-20(24(28-25)29-8-10-33-11-9-29)5-6-21(26-23)18-4-7-22(32-3)19(12-18)15-31/h4-7,12,16-17,31H,8-11,13-15H2,1-3H3/t16-,17+

InChI key

RFSMUFRPPYDYRD-CALCHBBNSA-N

General description

A cell-permeable pyrido-pyrimidine compound that acts as a highly selective and potent inhibitor against both mTORC1 & mTORC2 (IC50 = ~10 nM against mTOR activity in raptor or rictor immunoprecipitate from HEK293 with 100 µM ATP), with much reduced potency against MKK1 and PI 3-Kα (IC50 ~9 µM) and little or no activity toward more than 200 other protein and lipid kinases, including Akt, ATR, DNA-PK, PDK1, and PI 3-Kβ/γ/δ. Shown to effectively inhibit cellular mTOR phosphorylation on Ser2448/2481 as well as the phosphorylation and activation of mTORC1/2 downstream effector molecules such as Akt, S6K, 4E-BP1, but not RSK or Erk1/2, kinases that are not regulated by mTOR. Although Ku-63794 is shown not to affect IGF-1-induced Akt phosphorylation on Ser308 by PDK1 in mTORC2 knockout MEF cultures, Ku-63794 treatment of mTORC2-expressing MEF cells is shown to inhibit IGF-1-induced Akt Ser308 phosphorylation.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Malagu, K., et al. 2009. Bioorg. Med. Chem. Lett.19, 5950.
Garcia-Martinez, J.M., et al. 2009. Biochem. J.421, 29.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service