Skip to Content
Merck
All Photos(1)

Key Documents

4750-OP

Millipore

Glycerol

Omnipur®

Synonym(s):

Glycerol, 1,2,3-Propanetriol, Glycerine, 1,2,3-Propanetriol, Glycerin

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€78.30
4 L
€248.00

€78.30


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 L
€78.30
4 L
€248.00

About This Item

Linear Formula:
HOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
92.09
Beilstein:
635685
MDL number:
UNSPSC Code:
50161901
NACRES:
NA.28
technique(s):
electrophoresis: suitable
bp:
182 °C/20 mmHg (lit.)
vapor pressure:
<1 mmHg ( 20 °C)

€78.30


Estimated to ship on28 May 2025


Request a Bulk Order

vapor density

3.1 (vs air)

Quality Level

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥99%

form

viscous liquid

autoignition temp.

698 °F

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

technique(s)

electrophoresis: suitable

color

colorless

refractive index

n20/D 1.474 (lit.)

pH

~5 (20 °C, 100 g/L)

bp

182 °C/20 mmHg (lit.)

mp

20 °C (lit.)

density

1.25 g/mL (lit.)

shipped in

ambient

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Molecular Biology Grade.

A260 (0.05 M, water): ≤0.07 AU
A280 (0.05 M, water): ≤0.02 AU
Assay: ≥99%
Density at 25°C: 1.260-1.263 g/ml
DNAse: none detected
Identification: To pass test
Phosphatase: none detected
Protease: none detected
RNAse: none detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Warning

Toxicity: Irritant (B)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

390.2 °F - Pensky-Martens closed cup

Flash Point(C)

199 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service