Skip to Content
Merck
All Photos(2)

Key Documents

454870

Sigma-Aldrich

MARK/Par-1 Activity Inhibitor, 39621

The MARK/Par-1 Activity Inhibitor, 39621 controls the biological activity of MARK/Par-1. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

MARK/Par-1 Activity Inhibitor, 39621, Microtubule Affinity Regulating Kinase Inhibitor, MAP Affinity Regulating Kinase Inhibitor, PAR1 Inhibitor, N-(2,5-Dimethylphenyl)-2-(4-(4-methoxyphenyl)-3-oxo-3,4-dihydropyrazin-2-ylthio)acetamide, N-(2,5-Dimethylphenyl)-2-(4-(4-methoxyphenyl)-3-oxo-3,4-dihydropyrazin-2-ylthio)acetamide, Microtubule Affinity Regulating Kinase Inhibitor, MAP Affinity Regulating Kinase Inhibitor, PAR1 Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€250.00

€250.00


Estimated to ship on25 May 2025



Select a Size

Change View
10 MG
€250.00

About This Item

Empirical Formula (Hill Notation):
C21H21N3O3S
CAS Number:
Molecular Weight:
395.47
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€250.00


Estimated to ship on25 May 2025


Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige

solubility

DMSO: 100 mg/mL, clear, yellow

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C1C(SCC(NC2=C(C)C=CC(C)=C2)=O)=NC=CN1C3=CC=C(C=C3)OC

InChI

1S/C21H21N3O3S/c1-14-4-5-15(2)18(12-14)23-19(25)13-28-20-21(26)24(11-10-22-20)16-6-8-17(27-3)9-7-16/h4-12H,13H2,1-3H3,(H,23,25)

InChI key

CGRJEWQUCBVSJR-UHFFFAOYSA-N

General description

A cell-permeable dihydropyrazinyl-thioacetamide that acts as an ATP-competitive MARK inhibitor (IC50 = 3.6 µM; [ATP] = 100 µM) with much reduced or little activity against GSK-3β, SAD-kinase B/BRSK-1, MARKK/TAO-1, Cdc2/cycB, and p38/SAPK (30%, 20%, 11%, 4%, and 0% inhibition, respectively; [inhibitor] = 50 µM). Shown to effectively block primary rat cortical neuron axon growth (5.7 µm/hr vs 32.5 µm/hr with or without 20 µM inhibitor) and prevent MARK2 overexpression-induced Tau Ser262 phosphorylation and cytotoxicity due to microtubule network breakdown in CHO cells (10 µM).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Timm, T., et al. 2011. J. Biol. Chem.286, 41711.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anastasia Mashukova et al.
Molecular biology of the cell, 32(8), 690-702 (2021-02-18)
Par1b/MARK2 is a Ser/Thr kinase with pleiotropic effects that participates in the generation of apico-basal polarity in Caenorhabditis elegans. It is phosphorylated by atypical PKC(ι/λ) in Thr595 and inhibited. Because previous work showed a decrease in atypical protein kinase C
Zarin Tabassum et al.
The Journal of biological chemistry, 298(6), 101977-101977 (2022-04-27)
The posttranslational regulation of the neuronal proteome is critical for brain homeostasis but becomes dysregulated in the aged or diseased brain, in which abnormal posttranslational modifications (PTMs) are frequently observed. While the full extent of modified substrates that comprise the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service