Skip to Content
Merck
All Photos(1)

Key Documents

454850-M

Sigma-Aldrich

MAP Kinase 2, Mouse, Recombinant, E. coli

Synonym(s):

ERK2, MAPK2, p42 MAPK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
Pricing and availability is not currently available.

recombinant

expressed in E. coli

Quality Level

Assay

≥90% (SDS-PAGE)

form

liquid

specific activity

≥5000 units/mL

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

foreign activity

MEK, phosphatase, protease, none detected

storage temp.

−70°C

General description

Recombinant, mouse MAP Kinase 2 expressed in E. coli. A serine/threonine protein kinase that acts at the end of a protein kinase cascade linking signals from cell surface receptor tyrosine kinases to cytoplasmic and nuclear events. This protein was co-expressed with MEK and is phosphorylated and active.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will catalyze the transfer of 1.0 pmol phosphate to myelin basic protein per min at 30°, pH 7.5.

Physical form

In 100 mM NaCl, 50 mM HEPES, 1 mM DTT, 100 µM Na₂-EDTA, 50% glycerol, 0.01% Brij® 35 detergent, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Fukunaga, K., and Miyamoto, E. 1998. Mol. Neurobiol. 16, 79.
Wu, J., et al. 1993. Mol. Cell. Biol.13, 4539.

Legal Information

Brij is a registered trademark of Croda International PLC
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service