444898
Mastoparan
A cell-permeable synthetic peptide with sequence identical to Vespula lewisii.
Synonym(s):
Mastoparan, INLKALAALAKKIL
Select a Size
€200.00
Estimated to ship on27 May 2025
Select a Size
About This Item
€200.00
Estimated to ship on27 May 2025
Recommended Products
Quality Level
Assay
≥97% (HPLC)
form
solid
potency
7.5 μM IC50
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
white
solubility
water: 5 mg/mL
shipped in
ambient
storage temp.
−20°C
SMILES string
N([C@@H](C(CC)C)C(=O)N[C@@H](CC(C)C)C(=O)N)C(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](N)[C@H](CC)C)CC(=O)N)CC(C)C)CCCCN)C)CC(C)C)C)C)CC(C)C)C)C
InChI
1S/C70H131N19O15/c1-17-40(11)55(75)69(103)88-53(35-54(74)90)68(102)87-52(34-39(9)10)67(101)83-46(25-19-22-28-71)62(96)78-45(16)61(95)86-50(32-37(5)6)65(99)79-42(13)58(92)77-43(14)60(94)85-51(33-38(7)8)66(100)80-44(15)59(93)81-47(26-20-23-29-72)63(97)82-48(27-21-24-30-73)64(98)89-56(41(12)18-2)70(104)84-49(57(76)91)31-36(3)4/h36-53,55-56H,17-35,71-73,75H2,1-16H3,(H2,74,90)(H2,76,91)(H,77,92)(H,78,96)(H,79,99)(H,80,100)(H,81,93)(H,82,97)(H,83,101)(H,84,104)(H,85,94)(H,86,95)(H,87,102)(H,88,103)(H,89,98)/t40-,41?,42-,43-,44-,45-,46-,47-,48-,49-,50-,51-,52-,53-,55-,56-/m0/s1
InChI key
MASXKPLGZRMBJF-HDSAQESISA-N
General description
Biochem/physiol Actions
Na+,K+-ATPase activity
Packaging
Warning
Sequence
Physical form
Reconstitution
Other Notes
Liz, S.Z., et al. 1997. Brain Res.771, 184.
Klinker, J.F., et al. 1996. Biochem. Pharmacol. 51, 217.
Langel, U., et al. 1996. Regul. Peptides 62, 47.
Pfeiffer, D.R., et al. 1995. J. Biol. Chem. 270, 4923.
Igarashi, M., et al. 1993. Science259, 77.
Komatsu, M., et al. 1992. Endocrinology130, 221.
Adolfo Garcia-Sainz, J., et al. 1991. Biochem. Biophys. Res. Commun.179, 852.
Gil, J., et al. 1991. J. Cell Biol. 113, 943.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service