Skip to Content
Merck
All Photos(1)

Key Documents

444805

Sigma-Aldrich

Membrane Traffic Inhibitor, A5

The Membrane Traffic Inhibitor, A5 controls the biological activity of adaptor complex AP-1.

Synonym(s):

Membrane Traffic Inhibitor, A5, 1-(4-(4-(E-2-Methyl-2-butenyl)piperazinyl)phenyl)ethanone, HCl, 5431942, A5

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€205.00

€205.00


Estimated to ship on22 May 2025



Select a Size

Change View
10 MG
€205.00

About This Item

Empirical Formula (Hill Notation):
C17H24N2O · xHCl
Molecular Weight:
272.39 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

€205.00


Estimated to ship on22 May 2025


Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

pale yellow

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable piperazinyl compound that specifically blocks clathrin adaptor complex AP-1-dependent traffic between trans-Golgi network (TGN) and endosomes in budding yeast, while exhibiting little effects against other trafficking pathways involving TGN and endosomes. Inhibits growth of yeast cells lacking complementary GGA1/2 adaptor proteins (EC50 = 50 µM), but not wild-type or AP-1-deficient cells even at concentrations as high as 300 µM. A5 presumably acts at a stage after AP-1 membrane recruitment as evidenced by an enhanced AP-1 perinuclear localization upon A5 treatment of HeLa cells.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Duncan, M.C., et al. 2007. Proc. Natl. Acad. Sci. USA104, 6235.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service