Skip to Content
Merck
All Photos(1)

Key Documents

444200

Sigma-Aldrich

MDL-12,330A, Hydrochloride

Cell-permeable and irreversible inhibitor of adenylate cyclase (IC₅₀ = 250 µM).

Synonym(s):

MDL-12,330A, Hydrochloride, cis-N-(2-Phenylcyclopentyl)azacyclotridec-1-en-2-amine, HCl, RMI 12330

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€71.50

€71.50


Estimated to ship on16 April 2025



Select a Size

Change View
1 MG
€71.50

About This Item

Empirical Formula (Hill Notation):
C23H36N2 · xHCl
CAS Number:
Molecular Weight:
340.55 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€71.50


Estimated to ship on16 April 2025


Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL
water: 3 mg/mL
ethanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C23H36N2.ClH/c1-2-4-6-11-18-23(24-19-12-7-5-3-1)25-22-17-13-16-21(22)20-14-9-8-10-15-20;/h8-10,14-15,21-22H,1-7,11-13,16-19H2,(H,24,25);1H/t21-,22-;/m0./s1

InChI key

CKOPQUCSDBVAQG-VROPFNGYSA-N

General description

Cell-permeable and irreversible inhibitor of adenylate cyclase (IC50 = 250 µM).
Cell-permeable irreversible inhibitor of adenylate cyclase (IC50 = 250 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Adenylate cyclase
Product does not compete with ATP.
Reversible: no
Target IC50: 250 µM against adenylate cyclase

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 months at -20°C.

Other Notes

Correia-de-Sa, P., and Ribeiro, J.A. 1994. Eur. J. Pharmacol. 271, 349.
Gorczynska, E., et al. 1994. Endocrinology134, 293.
Delpiano, M.A., and Acker, H. 1991. J. Neurochem.57, 291.
Lippe, C., and Ardizzone, C. 1991. Comp. Biochem. Physiol.99, 209.
Guellaen, G., et al. 1977. Biochim. Biophys. Acta 484, 465.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service