Skip to Content
Merck
All Photos(1)

Key Documents

444105

Sigma-Aldrich

Anti-Mannose 6-Phosphate Receptor Mouse mAb (2G11)

liquid, clone 2G11, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€459.00

€459.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€459.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€459.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2G11, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

bovine, monkey, human

should not react with

hamster

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2a

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... M6PR(4074)

General description

Purified mouse monoclonal antibody. Recognizes the mannose 6-phosphate receptor protein.
Recognizes the ~300 kDa mannose 6-phosphate receptor protein.
This Anti-Mannose 6-Phosphate Receptor Mouse mAb (2G11) is validated for use in Immunoblotting, IF, IHC, IP for the detection of Mannose 6-Phosphate Receptor.

Immunogen

Bovine
Epitope: within the extracelluar domain
native bovine cation independent (CI)-mannose 6-phosphate receptor

Application

Immunoblotting (1 µg/ml, non-reducing, see comments)

Immunofluorescence (see comments)

Immunohistochemistry (2-5 µg/ml)

Immunoprecipitation (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Dintzis, S., et al. 1994. J. Biol. Chem.269, 12159.
Dintzis, S., and Pfeffer, S. 1990. EMBO J.9, 77.
Does not detect MPR in hamster (CHO) cells. Not suitable for immunoblotting using reducing conditions. This antibody has also been reported to work for immunoprecipitation and immunofluorescence. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer Hirst et al.
Human molecular genetics, 24(17), 4984-4996 (2015-06-19)
Adaptor proteins (AP 1-5) are heterotetrameric complexes that facilitate specialized cargo sorting in vesicular-mediated trafficking. Mutations in AP5Z1, encoding a subunit of the AP-5 complex, have been reported to cause hereditary spastic paraplegia (HSP), although their impact at the cellular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service