Skip to Content
Merck
All Photos(1)

Documents

437694

Sigma-Aldrich

DL-α-Lipoic Acid, Dihydro-

An effective antioxidant.

Synonym(s):

DL-α-Lipoic Acid, Dihydro-, DHLA, Dihydrolipoic Acid, Thiocytic Acid, Dihydro-

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16O2S2
CAS Number:
Molecular Weight:
208.34
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77

Quality Level

Assay

≥95% (TLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow

solubility

ethanol: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C8H16O2S2/c9-8(10)4-2-1-3-7(12)5-6-11/h7,11-12H,1-6H2,(H,9,10)

InChI key

IZFHEQBZOYJLPK-UHFFFAOYSA-N

General description

An effective antioxidant. Scavenges hypochlorous acid, peroxyl, superoxide, hydroxyl and nitric oxide radicals. Also exhibits prooxidant properties, as demonstrated by its ability to reduce iron ions and to generate reactive sulfur-containing radicals that can damage certain proteins, such as α1-antiproteinase and creatine kinase. DHLA has recently been shown to inhibit apoptosis in rat thymocytes induced by methylprednisolone and etoposide.
An effective antioxidant. Scavenges hypochlorous acid, peroxyl, superoxide, hydroxyl, and nitric oxide radicals. Also exhibits prooxidant properties, as demonstrated by its ability to reduce iron ions and to generate reactive sulfur-containing radicals that can damage certain proteins, such as α1-antiproteinase and creatine kinase. DHLA has recently been shown to inhibit apoptosis in rat thymocytes induced by methylprednisolone and etoposide.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Scavenges hypochlorous acid, peroxyl, superoxide, hydroxyl, and nitric oxide radicals
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Other Notes

Packer, L., et al. 1997. Adv. Pharmacol. 38, 79.
Bustamante, J., et al. 1995. Free Radic. Biol. Med. 19, 339.
Scott, B.C., et al. 1994. Free Radic. Res. 20, 119.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

348.1 °F - Cleveland open cup

Flash Point(C)

175.6 °C - Cleveland open cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service