Skip to Content
Merck
All Photos(1)

Key Documents

427218

Sigma-Aldrich

FX11

≥96% (HPLC), solid, lactate dehydrogenase A inhibitor, Calbiochem®

Synonym(s):

Lactate Dehydrogenase A Inhibitor, FX11

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€262.00

€262.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€262.00

About This Item

Empirical Formula (Hill Notation):
C22H22O4
CAS Number:
Molecular Weight:
350.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54

€262.00


Estimated to ship on21 May 2025


Request a Bulk Order

Product Name

Lactate Dehydrogenase A Inhibitor, FX11, Lactate Dehydrogenase A Inhibitor, FX11, CAS 213971-34-7, is a cell permeable, selective, reversible, NADH competitive inhibitor of lactate dehhydrogenase A (LDHA; Ki = 8 µM for human liver LDHA).

Quality Level

Assay

≥96% (HPLC)

form

solid

potency

8 μM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell permeable, gossypol analog that acts as a selective, reversible, and NADH competitive inhibitor of lactate dehhydrogenase A (LDHA; Ki = 8 µM for human liver LDHA). Does not affect the activities of LDHB and glyceraldehyde-3-phosphate dehydrogenase even at higher concentrations. Shown to increase oxygen consumption, reactive oxygen species production, and cell death in P493 human lymphoma B cells. Also shown to reduce mitochondrial transmembrane potential and ATP levels and increase AMP kinase activity. Exhibits synergistic toxicity in P493 cells when combined with FK866 (Cat. No. 481908). Blocks the progression of human lymphoma and pancreatic cancer xenografts and when used in combination with FK866 it induces lymphoma regression.

Application

Lactate Dehydrogenase A Inhibitor, FX11, CAS 213971-34-7, is a cell permeable, selective, reversible, NADH competitive inhibitor of lactate dehhydrogenase A (LDHA; Ki = 8 µM for human liver LDHA).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
LDHA

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Le, A., et al. 2011. Proc Natl Acad Sci USA.107, 2037.
Deck, L. M., et al. 1998. J Med Chem.41, 3879.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service