Skip to Content
Merck
All Photos(1)

Key Documents

420134

Sigma-Aldrich

JNK Inhibitor VII, TAT-TI-JIP153-163, Cell-Permeable

The JNK Inhibitor VII, TAT-TI-JIP153-163, Cell-Permeable controls the biological activity of JNK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

JNK Inhibitor VII, TAT-TI-JIP153-163, Cell-Permeable, H₂N-YGRKKRRQRRR-RPKRPTTLNLF-NH₂, TAT₃₈₋₄₈ -Truncated Inhibitor based on JNK-Interacting Protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
€259.00

€259.00


Estimated to ship on12 May 2025



Select a Size

Change View
2 MG
€259.00

About This Item

Empirical Formula (Hill Notation):
C125H220N52O27
Molecular Weight:
2883.42
UNSPSC Code:
12352200
NACRES:
NA.77

€259.00


Estimated to ship on12 May 2025


Quality Level

Assay

≥97% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

solubility

water: 1 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

The non-permeant JNK inhibitor peptide TI-JIP153-163 (Cat. No. 420133) is made cell-permeable with an N-terminal TAT protein transduction domain sequence. Shown to offer neuroprotection in a rat ischemic brain injury model in in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
JNK
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H-Tyr-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-Arg-Pro-Lys-Arg-Pro-Thr-Thr-Leu-Asn-Leu-Phe-NH₂

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Guan, Q.H., et al. 2006. Neuroscience, 139, 609.
Barr, R.K., et al. 2004. J. Biol. Chem.279, 36327.
Bogoyevitch, M.A., et al. 2003. Cell. Mol. Biol. Lett.8, 550.
Barr, R.K., et al. 2002. J. Biol. Chem.277, 10987.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service