Skip to Content
Merck
All Photos(1)

Key Documents

411543

Sigma-Aldrich

Mouse Anti-Human IgG₁, Fc Fragment Specific (HP6069) Biotin Conjugate

liquid, clone HP6069, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
€264.00

€264.00


Estimated to ship on29 May 2025



Select a Size

Change View
200 μG
€264.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.77

€264.00


Estimated to ship on29 May 2025


biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

secondary antibodies

clone

HP6069, monoclonal

form

liquid

does not contain

preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

ambient

storage temp.

−20°C

target post-translational modification

unmodified

General description

Purified mouse monoclonal antibody conjugated to biotin. Recognizes human IgG1 Fc fragment.
This Mouse Anti-Human IgG₁, Fc Fragment Specific (HP6069) Biotin Conjugate is validated for use in Enzyme Immunoassay for the detection of Human IgG₁, Fc Fragment Specific.

Application

Enzyme Immunoassay (1:500-1:2000)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Other Notes

Antibody should be titrated for optimal results in individual systems.



Cross-reactivity by ELISA against human myeloma proteins:

Human IgG1: 100%

Human IgG2: ≤0.1%

Human IgG3: ≤0.2%

Human IgG4: ≤0.1%

Human IgM: ≤0.1%

Human IgA: ≤0.1%

Human IgE: ≤0.1%



Cross-reactivity by ELISA against chimeric antibodies:

Human IgG1: 100%

Human IgG2: <0.02%

Human IgG3: <0.02%

Human IgG4: <0.02%

Human IgM: <0.02%

Human IgA: <0.01%

Human IgE: <0.001%

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Rivera-Correa et al.
eLife, 8 (2019-11-13)
Anemia is a common complication of malaria that is characterized by the loss of infected and uninfected erythrocytes. In mouse malaria models, clearance of uninfected erythrocytes is promoted by autoimmune anti-phosphatidylserine (PS) antibodies produced by T-bet+B-cells, which bind to exposed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service