Skip to Content
Merck
All Photos(2)

Key Documents

410960

Sigma-Aldrich

Ischemin

A cell-permeable azobenzene compound that reversibly targets CBP-BRD acetyl-lysine binding pocket with moderate selectivity over BAZ1B, PCAF, BRD4-1 and BAZ2B.

Synonym(s):

Ischemin, (E)-5-(2-Amino-4-hydroxy-5-methyl-phenylazo)-2,4-dimethyl-benzenesulfonic acid, MS120, BRD4 Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17N3O4S
CAS Number:
Molecular Weight:
335.38
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.51
Pricing and availability is not currently available.

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange-red

solubility

DMSO: 50 mg/mL, orange

shipped in

ambient

storage temp.

2-8°C

SMILES string

CC1=C(/N=N/C2=C(N)C=C(O)C(C)=C2)C=C(C(C)=C1)S(=O)(O)=O

InChI key

UUECJWRVDTUDCB-ISLYRVAYSA-N

General description

A cell-permeable azobenzene compound that reversibly targets CREB-binding protein bromodomain (CBP-BRD) acetyl-lysine binding pocket (Kd = 19 µM) with moderate selectivity over BAZ1B, PCAF, BRD4-1 and BAZ2B (Kd = 37, 41, 70 and >100 µM, respectively) and disrupts CBP interaction with the binding partners. Blocks CBP-p53 interaction and alters the post-translational modification states on p53 and histones. Shown to inhibit the transcription functions of p53 on DNA damage (IC50 = 5 µM for p53-induced p21 activation in Luc-U2OS cells) and suppress cardiac myocyte apoptosis during ischemia conditions.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Other Notes

Borah, J.C., et al. 2011, Chem. Biol.18, 531.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service