Skip to Content
Merck
All Photos(1)

Key Documents

407719

Sigma-Aldrich

Iodoacetic Acid, Sodium Salt

Inhibits cysteine proteases.

Synonym(s):

Iodoacetic Acid, Sodium Salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H2IO2 · Na
CAS Number:
Molecular Weight:
207.93
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white to pale yellow

solubility

water: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C2H3IO2.Na/c3-1-2(4)5;/h1H2,(H,4,5);/q;+1/p-1

InChI key

AGDSCTQQXMDDCV-UHFFFAOYSA-M

General description

Inhibits cysteine proteases.

Packaging

Packaged under inert gas

Warning

Toxicity: Toxic (F)

Reconstitution

Unstable in solution; reconstitute just prior to use.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

406.4 °F

Flash Point(C)

208 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service