Skip to Content
Merck
All Photos(1)

Key Documents

407293

Sigma-Aldrich

α-Interferon, Mouse, Recombinant, E. coli

Sign Into View Organizational & Contract Pricing

Select a Size

100 KU
€615.00

€615.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 KU
€615.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€615.00


Estimated to ship on25 May 2025


Request a Bulk Order

description

Merck USA index - 14, 4990

Quality Level

Assay

≥95% (SDS-PAGE)

form

liquid

specific activity

≥3.5 x 104 units/mL

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Recombinant, mouse α-interferon expressed in E. coli. Antiviral, antineoplastic agent. Expressed during the triggering stage of macrophage cytocidal activation. Suppresses bone marrow function and cell proliferation. Modulates the immune response. Also known to induce apoptotic cell death in non-melanoma skin cancer.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit of interferon per ml of medium will cause a 50% inhibition of EMCV-induced cytopathic effect on mouse L929 cells. Units are determined with respect to the NIH international reference standard for mouse IFN-α.

Other Notes

Krishnan, K., et al. 1997. Eur. J. Biochem. 247, 298.
Rodriquez-Villanueva, S., and McDonnel, T.J. 1995. Int. J. Cancer61, 110.
Dianzani, F. 1993. Gut34, 574.
Brouty-Boyce, D., and Zetter, B.R. 1990. Science208, 516.
Sidky, Y.A., and Borden, E.C. 1987. Cancer Res.47, 5155.
Daugherty, B.L., and Pestka, S. 1986. Methods Enzymol.119, 34.
Pestka, S. 1986. Methods Enzymol.119, 14.
Familletti, P.C., et al. 1981. Methods Enzymol.78, 387.
Johnson, M.D., et al. 1981. Methods Enzymol.78, 394.
Rubinstein, S., et al. 1981. J. Virol.37, 755.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Markland et al.
Antimicrobial agents and chemotherapy, 44(4), 859-866 (2000-03-18)
The enzyme IMP dehydrogenase (IMPDH) catalyzes an essential step in the de novo biosynthesis of guanine nucleotides, namely, the conversion of IMP to XMP. The major event occurring in cells exposed to competitive IMPDH inhibitors such as ribavirin or uncompetitive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service